1,3,5-Triisopropylbenzene ≥95.0%

Supplier: TCI America

T0458-500ML T0458-25ML T0458-100ML
TCT0458-500MLEA 290.72 CAD
TCT0458-500ML TCT0458-025ML TCT0458-100ML
1,3,5-Triisopropylbenzene ≥95.0%
1,3,5-Triisopropylbenzene

CAS Number: 717-74-8
MDL Number: MFCD00008890
Molecular Formula: C15H24
Molecular Weight: 204.36
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 235
Flash Point (°C): 86
Specific Gravity (20/20): 0.85

Formula: C₁₅H₂₄
Boiling Pt: 235 °C
Melting Pt: –7 °C
Density: 0.85 (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00008890
CAS Number: 717-74-8
EINECS: 211-941-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f382e17a04a2858d9f799ddadd3a44cbd44dae58&vid=7c6d10a56428570697500eea6c693e4fe61b4ab4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR