2,4,6-Triiodophenol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T0452-25G T0452-5G
TCT0452-025GEA 418.7 CAD
TCT0452-025G TCT0452-5G
2,4,6-Triiodophenol ≥98.0% (by HPLC, titration analysis)
2,4,6-Triiodophenol

CAS Number: 609-23-4
MDL Number: MFCD00002179
Molecular Formula: C6H3I3O
Molecular Weight: 471.80
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 160

Formula: C₆H₃I₃O
MW: 471.8 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 160 °C
MDL Number: MFCD00002179
CAS Number: 609-23-4
EINECS: 210-186-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80a5977f87a2d306d9da67e7a77f940f1c09faf8&vid=cfb00da2206320c1d5c02f66e9596c86148bd0ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR