2,3,5-Triiodobenzoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
T0451-25G
T0451-5G
TCT0451-025GEA
201.61
CAD
TCT0451-025G
TCT0451-005G
2,3,5-Triiodobenzoic acid ≥98.0% (by HPLC, titration analysis)
2,3,5-Triiodobenzoic acid
CAS Number: 88-82-4
MDL Number: MFCD00002420
Molecular Formula: C7H3I3O2
Molecular Weight: 499.81
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 231
Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: TIBA
Formula:
C₇H₃I₃O₂ Boiling Pt: 416 °C (1013 hPa) Melting Pt: 231 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002420 CAS Number: 88-82-4 EINECS: 201-859-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...