α,α,α-Trifluoro-m-cresol ≥98.0%

Supplier: TCI America
Warning

T0436-250G T0436-25G
TCT0436-250GEA 317.42 CAD
TCT0436-250G TCT0436-025G
α,α,α-Trifluoro-m-cresol ≥98.0%
α,α,α-Trifluoro-m-cresol

CAS Number: 98-17-9
MDL Number: MFCD00002299
Molecular Formula: C7H5F3O
Molecular Weight: 162.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 178
Flash Point (°C): 73
Specific Gravity (20/20): 1.36


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Hydroxy-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-Trifluoro-m-cresol, 3-(Trifluoromethyl)phenol

Formula: C₇H₅F₃O
Boiling Pt: 178 °C
Melting Pt: –2 °C
Density: 1.36 (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002299
CAS Number: 98-17-9
EINECS: 202-645-5
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52dd503e05c90fe28f8aeff87d886c29ec5c885b&vid=52dd503e05c90fe28f8aeff87d886c29ec5c885b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR