Triethylene glycol ≥99.0%

Supplier: TCI America

Synonyms: 2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol

T0428-25G T0428-500G
TCT0428-025GEA 37.45 CAD
TCT0428-025G TCT0428-500G
Triethylene glycol ≥99.0%
Triethylene glycol

CAS Number: 112-27-6
MDL Number: MFCD00002880
Molecular Formula: C6H14O4
Molecular Weight: 150.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 276
Flash Point (°C): 174
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: Triglycol

Formula: C₆H₁₄O₄
MW: 150.17 g/mol
Boiling Pt: 276 °C
Melting Pt: –7 °C
Density: 1.13 (20 °C)
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00002880
CAS Number: 112-27-6
EINECS: 203-953-2
Merck Index: 12,09802
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=077e60635e9f31a1bf9a36de51860812a4eb8769&vid=077e60635e9f31a1bf9a36de51860812a4eb8769&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR