Triethylammonium chloride ≥98.5%

Supplier: TCI America
Warning

Synonyms: Triethylamine hydrochloride

T0425-500G T0425-25G
TCT0425-500GEA 170.64 CAD
TCT0425-500G TCT0425-025G
Triethylammonium chloride ≥98.5%
Triethylammonium chloride

CAS Number: 554-68-7
MDL Number: MFCD00012500
Molecular Formula: C6H15N
Molecular Weight: 137.65
Purity/Analysis Method: >98.5% (T)
Form: Crystal
Color: White
Melting point (°C): 258

Formula: C₆H₁₆ClN
MW: 137.65 g/mol
Melting Pt: 258 °C
Density: 0.7637 g/cm³ (20 °C)
MDL Number: MFCD00012500
CAS Number: 554-68-7
EINECS: 209-067-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffe30da07f588ee28d9787be79b97beaaeae1274&vid=ffe30da07f588ee28d9787be79b97beaaeae1274&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR