Tributyl phosphite ≥95.0%

Supplier: TCI America

Synonyms: Tri-n-butylphosphite

T0362-500ML T0362-25ML
TCT0362-500MLEA 216.93 CAD
TCT0362-500ML TCT0362-25ML
Tributyl phosphite ≥95.0%
Tributyl phosphite

CAS Number: 102-85-2
MDL Number: MFCD00009437
Molecular Formula: C12H27O3P
Molecular Weight: 250.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 125
Melting point (°C): -80
Flash Point (°C): 129
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Phosphorous Acid Tributyl Ester

Formula: C₁₂H₂₇O₃P
MW: 250.32 g/mol
Boiling Pt: 125 °C
Melting Pt: –80 °C
Density: 0.92 (20 °C)
Flash Pt: 129 °C
MDL Number: MFCD00009437
CAS Number: 102-85-2
EINECS: 203-061-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d222230e1915e9ea51cfe511ca8f2fa1485542f1&vid=d222230e1915e9ea51cfe511ca8f2fa1485542f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR