1,3,5-Tribromobenzene ≥98.0%
Supplier: TCI America
T0347-25G
T0347-5G
T0347-250G
TCT0347-025GEA
67.5
CAD
TCT0347-025G
TCT0347-005G
TCT0347-250G
1,3,5-Tribromobenzene ≥98.0%
1,3,5-Tribromobenzene
CAS Number: 626-39-1
MDL Number: MFCD00000080
Molecular Formula: C6H3Br3
Molecular Weight: 314.80
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 271
Melting point (°C): 122
Formula:
C₆H₃Br₃ Boiling Pt: 271 °C Melting Pt: 122 °C Density: 2.637 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000080 CAS Number: 626-39-1 EINECS: 210-947-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...