1,2,4-Tribromobenzene ≥95.0%
Supplier: TCI America
|
Warning
|
T0346-25G
T0346-5G
TCT0346-025GEA
243.32
CAD
TCT0346-025G
76220-484
1,2,4-Tribromobenzene ≥95.0%
1,2,4-Tribromobenzene
CAS Number: 615-54-3
MDL Number: MFCD00000062
Molecular Formula: C6H3Br3
Molecular Weight: 314.80
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 42
Flash Point (°C): 113
Formula:
C₆H₃Br₃ Boiling Pt: 85…90 °C (0.04 mmHg) Melting Pt: 42 °C Flash Pt: 113 °C |
MDL Number:
MFCD00000062 CAS Number: 615-54-3 EINECS: 210-433-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...