2-(4-Toluoyl)benzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(4-Methylbenzoyl)benzenecarboxylic acid

T0308-500G T0308-25G
TCT0308-500GEA 393.42 CAD
TCT0308-500G TCT0308-025G
2-(4-Toluoyl)benzoic acid ≥98.0% (by titrimetric analysis)
2-(4-Toluoyl)benzoic acid

CAS Number: 85-55-2
MDL Number: MFCD00020287
Molecular Formula: C15H12O3
Molecular Weight: 240.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 138


Other Product Names / Synonyms: 4-Methylbenzophenone-2'-carboxylic Acid, 2-(4-Methylbenzoyl)benzoic Acid, p-Toluoyl-o-benzoic Acid

Formula: C₁₅H₁₂O₃
MW: 240.26 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00020287
CAS Number: 85-55-2
EINECS: 201-614-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfd0cd730f174e76ce5534974e6ef3b623f04476&vid=cfd0cd730f174e76ce5534974e6ef3b623f04476&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR