m-Toluonitrile

Supplier: TCI America

Synonyms: m-Tolunitrile, m-Methylbenzonitrile, 3-Methylbenzonitrile

T0305-25ML T0305-500ML
TCT0305-025MLEA 26.86 CAD
TCT0305-025ML TCT0305-500ML
m-Toluonitrile
m-Toluonitrile

CAS Number: 620-22-4
MDL Number: MFCD00001808
Molecular Formula: C8H7N
Molecular Weight: 117.15
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 210
Flash Point (°C): 86
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: 3-Cyanotoluene

Formula: C₈H₇N
Boiling Pt: 210 °C
Melting Pt: –0.5 °C
Density: 0.99 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00001808
CAS Number: 620-22-4
EINECS: 210-631-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc5aeef5485d4e301f0a57f0cbd3336dea37b40a&vid=cc5aeef5485d4e301f0a57f0cbd3336dea37b40a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR