Methyl-p-toluate ≥98.0%
Supplier: TCI America
|
Warning
|
T0297-25G
T0297-500G
TCT0297-025GEA
26.86
CAD
TCT0297-025G
TCT0297-500G
Methyl-p-toluate ≥98.0%
Methyl p-toluate
CAS Number: 99-75-2
MDL Number: MFCD00008441
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 223
Flash Point (°C): 98
Freezing point (°C): 32
Other Product Names / Synonyms: 4-Methylbenzoic Acid Methyl Ester, Methyl 4-Methylbenzoate, p-Toluic Acid Methyl Ester
Formula:
C₉H₁₀O₂ Boiling Pt: 223 °C Melting Pt: 33…34 °C Density: 1.06 g/cm³ (20 °C) Flash Pt: 98 °C |
MDL Number:
MFCD00008441 CAS Number: 99-75-2 EINECS: 202-784-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...