Methyl-o-toluate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 2-methylbenzoate, o-Toluic acid methyl ester, 2-Toluic acid methyl ester, Methyl-2-methylbenzoate, 2-Methylbenzoic acid methyl ester
CAS Number: 89-71-4
MDL Number: MFCD00008428
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 213
Flash Point (°C): 82
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 2-Methylbenzoic Acid Methyl Ester, o-Toluic Acid Methyl Ester
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 213 °C Melting Pt: –50 °C Density: 1.07 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00008428 CAS Number: 89-71-4 EINECS: 201-932-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...