Ethyl-O-toluate ≥98.0%

Supplier: TCI America

T0294-25G T0294-500G
TCT0294-025GEA 61.62 CAD
TCT0294-025G TCT0294-500G
Ethyl-O-toluate ≥98.0%
Ethyl o-toluate

CAS Number: 87-24-1
MDL Number: MFCD00009112
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 227
Flash Point (°C): 91
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Ethyl 2-Methylbenzoate, 2-Methylbenzoic Acid Ethyl Ester, o-Toluic Acid Ethyl Ester

Formula: C₁₀H₁₂O₂
Boiling Pt: 227 °C
Melting Pt: –10 °C
Density: 1.04 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00009112
CAS Number: 87-24-1
EINECS: 201-734-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da35a4e1062c0611ad8b33ed1ed8c4d136eba647&vid=da35a4e1062c0611ad8b33ed1ed8c4d136eba647&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR