p-Toluic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
4-Methylbenzoic acid, 4-Toluic acid
T0293-25G
T0293-500G
TCT0293-025GEA
44.26
CAD
TCT0293-025G
TCT0293-500G
p-Toluic acid ≥98.0%
p-Toluic acid
CAS Number: 99-94-5
MDL Number: MFCD00002565
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 275
Melting point (°C): 181
Flash Point (°C): 181
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Methylbenzoic Acid
Formula:
C₈H₈O₂ Boiling Pt: 275 °C Melting Pt: 181 °C Density: 1.06 g/cm³ (20 °C) Flash Pt: 181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002565 CAS Number: 99-94-5 EINECS: 202-803-3 Merck Index: 12,09673 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...