Toluene-2-sulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 2-Methylbenzene-1-sulfonamide, 2-Methylbenzenesulfonamide, o-Toluenesulfonamide, 2-Methylbenzene-1-sulphonamide, 2-Methylbenzenesulphonamide, o-Toluenesulphonamide

T0280-25G T0280-500G
TCT0280-025GEA 69.14 CAD
TCT0280-025G TCT0280-500G
Toluene-2-sulfonamide ≥98.0% (by HPLC, total nitrogen)
Toluene-2-sulphonamide

CAS Number: 88-19-7
MDL Number: MFCD00007934
Molecular Formula: C7H9NO2S
Molecular Weight: 171.21
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 155

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00007934
CAS Number: 88-19-7
EINECS: 201-808-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d76beb6168b92d38ecac0e7978edb4d1f62f2c6&vid=8d76beb6168b92d38ecac0e7978edb4d1f62f2c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR