Ethyl toluene-4-sulfonate ≥98.0%

Supplier: TCI America

T0268-25G T0268-500G
TCT0268-025GEA 39.5 CAD
TCT0268-025G TCT0268-500G
Ethyl toluene-4-sulfonate ≥98.0%
Ethyl toluene-4-sulphonate

CAS Number: 80-40-0
MDL Number: MFCD00009100
Molecular Formula: C9H12O3S
Molecular Weight: 200.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 173
Flash Point (°C): 157
Freezing point (°C): 30


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: p-Toluenesulfonic Acid Ethyl Ester, Ethyl Tosylate

Formula: C₉H₁₂O₃S
Boiling Pt: 173 °C
Melting Pt: 30…32 °C
Density: 1.174 g/cm³ (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00009100
CAS Number: 80-40-0
EINECS: 201-276-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,03889
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d3529de5c2dfb99c83b8e8acf95170de734a47&vid=25d3529de5c2dfb99c83b8e8acf95170de734a47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR