Toluene-3,4-dithiol ≥97.0%
Supplier: TCI America
Synonyms:
3,4-Toluenedithiol, 4-Methyl-1,2-dimercaptobenzene, 1,2-Dimercapto-4-methylbenzene, 4-Methylbenzene-1,2-dithiol
CAS Number: 496-74-2
MDL Number: MFCD00004844
Molecular Formula: C7H8S2
Molecular Weight: 156.26
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 137
Flash Point (°C): 113
Freezing point (°C): 30
Product Categories: Synthetic Organic Chemistry, Thiols, Materials Chemistry, Molecular Conductors, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: Dithiol
Formula:
C₇H₈S₂ Boiling Pt: 137 °C Melting Pt: 30…32 °C Density: 1.18 g/cm³ (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00004844 CAS Number: 496-74-2 EINECS: 207-828-3 Merck Index: 12,09669 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...