4-Tolualdehyde ≥98.0%

Supplier: TCI America
Warning

T0259-500ML T0259-25ML T0259-100ML
TCT0259-500MLEA 281.18 CAD
TCT0259-500ML TCT0259-025ML TCT0259-100ML
4-Tolualdehyde ≥98.0%
4-Tolualdehyde

CAS Number: 104-87-0
MDL Number: MFCD00006954
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 204
Flash Point (°C): 88
Specific Gravity (20/20): 1.02


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction), Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Methylbenzaldehyde

Formula: C₈H₈O
Boiling Pt: 204 °C
Melting Pt: –6 °C
Density: 1.02 (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00006954
CAS Number: 104-87-0
EINECS: 203-246-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3615c889da76777e498fec283149fcd93236263a&vid=13ce3ff8201f4845973f4192f9e0072aef0435b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR