3-Tolualdehyde ≥97.0% stabilized
Supplier: TCI America
Synonyms:
m-Tolualdehyde, 3-Methylbenzaldehyde, m-Tolylaldehyde
Stabilization:
Stabilised with Stabilised
T0258-25ML
T0258-5ML
TCT0258-025MLEA
107.44
CAD
TCT0258-025ML
TCT0258-5ML
3-Tolualdehyde ≥97.0% stabilized
3-Tolualdehyde
(stabilized with HQ)
CAS Number: 620-23-5
MDL Number: MFCD00003374
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 200
Flash Point (°C): 78
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: 3-Methylbenzaldehyde
Formula:
C₈H₈O MW: 120.15 g/mol Boiling Pt: 200 °C Density: 1.03 (20 °C) Flash Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003374 CAS Number: 620-23-5 EINECS: 210-632-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...