DL-α-Tocopherol acetate ≥98.0%

Supplier: TCI America

Synonyms: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate

T0252-500G T0252-25G T0252-100G
TCT0252-500GEA 426.6 CAD
TCT0252-500G TCT0252-025G TCT0252-100G
DL-α-Tocopherol acetate ≥98.0%
DL-α-Tocopherol acetate

CAS Number: 7695-91-2
MDL Number: MFCD00072042
Molecular Formula: C31H52O3
Molecular Weight: 472.75
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 224
Flash Point (°C): 110
Specific Gravity (20/20): 0.97


Product Categories: Bioscience, Antioxidants, Vitamins, Terpenes (Others)
Other Product Names / Synonyms: Vitamin E Acetate

Formula: C₃₁H₅₂O₃
Boiling Pt: 224 °C
Density: 0.97 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00072042
CAS Number: 7695-91-2
EINECS: 231-710-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae38089251a46b446d1028b56ae8e51e32fe1306&vid=ae38089251a46b446d1028b56ae8e51e32fe1306&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR