DL-α-Tocopherol (Vitamin E) ≥96.0%

Supplier: TCI America

T0251-250G T0251-25G
TCT0251-250GEA 284.48 CAD
TCT0251-250G TCT0251-025G
DL-α-Tocopherol (Vitamin E) ≥96.0%
DL-α-Tocopherol

CAS Number: 10191-41-0
MDL Number: MFCD00072051
Molecular Formula: C29H50O2
Molecular Weight: 430.72
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Yellow
Boiling point (°C): 220
Melting point (°C): 4
Flash Point (°C): 240
Specific Gravity (20/20): 0.95


Product Categories: Bioscience, Antioxidants, Vitamins, Terpenes (Others)
Other Product Names / Synonyms: Vitamin E

Formula: C₂₉H₅₀O₂
Boiling Pt: 220 °C
Melting Pt: 4 °C
Density: 0.95 (20 °C)
Flash Pt: 240 °C
MDL Number: MFCD00072051
CAS Number: 10191-41-0
EINECS: 233-466-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1d41817f60f683437f44d4c16cbcde040111774&vid=e1d41817f60f683437f44d4c16cbcde040111774&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR