Thymolphthalein

Supplier: TCI America

Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one

T0237-1G T0237-25G
TCT0237-001GEA 34.76 CAD
TCT0237-001G TCT0237-025G
Thymolphthalein
Thymolphthaleine

CAS Number: 125-20-2
MDL Number: MFCD00005909
Molecular Formula: C28H30O4
Molecular Weight: 430.54
Form: Crystal
Color: White
Melting point (°C): 253

Formula: C₂₈H₃₀O₄
MW: 430.54 g/mol
Melting Pt: 253 °C
Storage Temperature: Ambient
MDL Number: MFCD00005909
CAS Number: 125-20-2
EINECS: 204-729-7
Merck Index: 12,09542
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9684b17cfbd4fec22a37b3016ce13c12d9edc16f&vid=9684b17cfbd4fec22a37b3016ce13c12d9edc16f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR