Thymol blue

Supplier: TCI America

T0235-1G T0235-25G
TCT0235-001GEA 58.46 CAD
TCT0235-001G TCT0235-025G
Thymol blue
Thymol blue

CAS Number: 76-61-9
MDL Number: MFCD00005869
Molecular Formula: C27H30O5S
Molecular Weight: 466.59
Form: Crystal
Color: Yellow
Melting point (°C): 222


Other Product Names / Synonyms: Thymolsulfonphthalein

Formula: C₂₇H₃₀O₅S
Melting Pt: 222 °C
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71696df4ca58faa71150ac7d4962c48814711cb5&vid=71696df4ca58faa71150ac7d4962c48814711cb5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR