D-Threonine ≥98.0%
Supplier: TCI America
Synonyms:
H-D-Thr-OH, (2R,3S)-2-Amino-3-hydroxybutanoic acid
T0228-25G
T0228-500G
T0228-100G
TCT0228-025GEA
143.78
CAD
TCT0228-025G
TCT0228-500G
TCT0228-100G
D-Threonine ≥98.0%
D-Threonine
CAS Number: 632-20-2
MDL Number: MFCD00064269
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: 28 deg (C=3, H2O)
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Thr-OH
Formula:
C₄H₉NO₃ MW: 119.12 g/mol |
MDL Number:
MFCD00064269 CAS Number: 632-20-2 EINECS: 211-171-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...