2-Thenoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0218-25G T0218-100G
TCT0218-250GEA 421.5 CAD
TCT0218-250G TCT0218-025G TCT0218-100G
2-Thenoic acid ≥98.0% (by titrimetric analysis)
2-Thenoic acid

CAS Number: 527-72-0
MDL Number: MFCD00005437
Molecular Formula: C5H4O2S
Molecular Weight: 128.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 260
Melting point (°C): 128


Other Product Names / Synonyms: 2-Thenoic Acid

Formula: C₅H₄O₂S
Boiling Pt: 260 °C
Melting Pt: 128 °C
MDL Number: MFCD00005437
CAS Number: 527-72-0
EINECS: 208-423-4
Merck Index: 12,09491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33de5a2074a6b065870b7a5db77225ca755a9eb2&vid=33de5a2074a6b065870b7a5db77225ca755a9eb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR