3,3'-Thiodipropionic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,3-Thiodipropionic acid

T0204-500G T0204-25G
TCT0204-500GEA 56.25 CAD
TCT0204-500G TCT0204-025G
3,3'-Thiodipropionic acid ≥99.0% (by titrimetric analysis)
3,3'-Thiodipropionic acid

CAS Number: 111-17-1
MDL Number: MFCD00002781
Molecular Formula: C6H10O4S
Molecular Weight: 178.20
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 131
Flash Point (°C): 128

Formula: C₆H₁₀O₄S
MW: 178.21 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 131 °C
Density: 1.496 g/cm³ (20 °C)
Flash Pt: 128 °C
MDL Number: MFCD00002781
CAS Number: 111-17-1
EINECS: 203-841-3
Merck Index: 12,09468
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87b6116d1685dea3879fe51e45cd78fd4cb7201f&vid=87b6116d1685dea3879fe51e45cd78fd4cb7201f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR