Thiobenzoic acid ≥93.0% (by titrimetric analysis)

Supplier: TCI America

T0195-25G T0195-500G
TCT0195-025GEA 43.88 CAD
TCT0195-025G TCT0195-500G
Thiobenzoic acid ≥93.0% (by titrimetric analysis)
Thiobenzoic acid

CAS Number: 98-91-9
MDL Number: MFCD00004852
Molecular Formula: C7H6OS
Molecular Weight: 138.18
Purity/Analysis Method: >93.0% (T)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 87
Flash Point (°C): 94
Freezing point (°C): 20
Specific Gravity (20/20): 1.18
Storage Temperature: 0-10°C

Formula: C₇H₆OS
Boiling Pt: 87 °C
Melting Pt: 24 °C
Density: 1.18 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00004852
CAS Number: 98-91-9
EINECS: 202-712-9
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37921c4a42e7e02540ce1a204376c2a365d8c199&vid=37921c4a42e7e02540ce1a204376c2a365d8c199&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR