1,1,4,4-Tetraphenyl-1,3-butadiene ≥99.0% (by HPLC)

Supplier: TCI America
Warning

T0168-5G T0168-1G
TCT0168-005GEA 339.7 CAD
TCT0168-005G TCT0168-001G
1,1,4,4-Tetraphenyl-1,3-butadiene ≥99.0% (by HPLC)
1,1,4,4-Tetraphenyl-1,3-butadiene

CAS Number: 1450-63-1
MDL Number: MFCD00004766
Molecular Formula: C28H22
Molecular Weight: 358.48
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 201

Formula: (C₆H₅)₂CCHCHC(C₆H₅)₂
MW: 358.48 g/mol
Boiling Pt: 556 °C (1013 hPa)
Melting Pt: 201 °C
MDL Number: MFCD00004766
CAS Number: 1450-63-1
EINECS: 215-914-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df754df1e966048442992445dddf31ffdf4f4e04&vid=df754df1e966048442992445dddf31ffdf4f4e04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR