Tetraphenylethylene ≥98.0%
Supplier: TCI America
Synonyms:
1,1,2,2-Tetraphenylethylene
T0167-1G
T0167-10G
TCT0167-001GEA
79
CAD
TCT0167-001G
TCT0167-010G
Tetraphenylethylene ≥98.0%
Tetraphenylethylene
CAS Number: 632-51-9
MDL Number: MFCD00004764
Molecular Formula: C26H20
Molecular Weight: 332.45
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 225
Formula:
C₂₆H₂₀ Boiling Pt: 420 °C (1013 hPa) Melting Pt: 225 °C Density: 1.148 g/cm³ (20 °C) |
MDL Number:
MFCD00004764 CAS Number: 632-51-9 EINECS: 211-179-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...