Tetramethylurea ≥98.0%
Supplier: TCI America
|
Danger
|
T0158-25G
T0158-500G
TCT0158-025GEA
52.55
CAD
TCT0158-025G
TCT0158-500G
Tetramethylurea ≥98.0%
Tetramethylurea
CAS Number: 632-22-4
MDL Number: MFCD00008319
Molecular Formula: C5H12N2O
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 177
Melting point (°C): -1
Flash Point (°C): 75
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: TMU
Formula:
C₅H₁₂N₂O Boiling Pt: 177 °C Melting Pt: –1 °C Density: 0.97 (20 °C) Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008319 CAS Number: 632-22-4 EINECS: 211-173-9 Merck Index: 12,09368 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...