1,1,3,3-Tetramethyl-2-thiourea ≥98.0%

Supplier: TCI America

Synonyms: N,N,N',N'-Tetramethylthiourea, Tetramethyl-2-thiourea, Tetramethylthiourea

T0156-25G T0156-500G T0156-100G
TCT0156-025GEA 88.48 CAD
TCT0156-025G TCT0156-500G TCT0156-100G
1,1,3,3-Tetramethyl-2-thiourea ≥98.0%
1,1,3,3-Tetramethyl-2-thiourea

CAS Number: 2782-91-4
MDL Number: MFCD00008324
Molecular Formula: C5H12N2S
Molecular Weight: 132.23
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Boiling point (°C): 243
Melting point (°C): 80

Formula: C₅H₁₂N₂S
MW: 132.23 g/mol
Boiling Pt: 245
Melting Pt: 80 °C
MDL Number: MFCD00008324
CAS Number: 2782-91-4
EINECS: 220-488-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cefb4959c41be1ee2b726cb1901515c838b515e0&vid=cefb4959c41be1ee2b726cb1901515c838b515e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR