N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride ≥98.0%

Supplier: TCI America
Warning

Synonyms: N1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochloride, TMPD 2 HCl, N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochloride, N,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride

T0151-25G T0151-1G T0151-5G
TCT0151-025GEA 189.6 CAD
TCT0151-025G TCT0151-001G TCT0151-5G
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride ≥98.0%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride

CAS Number: 637-01-4
MDL Number: MFCD00012482
Molecular Formula: C10H16N2
Molecular Weight: 237.17
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 224


Other Product Names / Synonyms: TMPD

Formula: C₁₀H₁₈Cl₂N₂
MW: 237.17 g/mol
Melting Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD00012482
CAS Number: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41caff63a133ca84e4409814fc36fe408ff2e201&vid=41caff63a133ca84e4409814fc36fe408ff2e201&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR