Tetramethylammonium iodide

Supplier: TCI America

T0139-100G T0139-25G T0139-500G
TCT0139-100GEA 178.54 CAD
TCT0139-100G TCT0139-025G TCT0139-500G
Tetramethylammonium iodide
Tetramethylammonium iodide

CAS Number: 75-58-1
MDL Number: MFCD00011629
Molecular Formula: C4H12IN
Molecular Weight: 201.05
Form: Crystal
Color: White

Formula: C₄H₁₂IN
MW: 201.05 g/mol
Melting Pt: 300
Density: 1.84 g/cm³ (20 °C)
MDL Number: MFCD00011629
CAS Number: 75-58-1
EINECS: 200-881-3
UN: 2811
ADR: 6.1 III
Merck Index: 12,09364
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6433c74b5173d5c63e3b36be80568b85a5d0459b&vid=6433c74b5173d5c63e3b36be80568b85a5d0459b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR