1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) ≥98.0%
Supplier: TCI America
T0134-500G
T0134-25G
TCT0134-500GEA
242.7
CAD
TCT0134-500G
TCT0134-025G
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) ≥98.0%
1,2,3,4-Tetrahydronaphthalen-1-one (1-Tetralone, α-Tetralone)
CAS Number: 529-34-0
MDL Number: MFCD00001688
Molecular Formula: C10H10O
Molecular Weight: 146.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 257
Flash Point (°C): 129
Freezing point (°C): 7
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: 3,4-Dihydro-1(2H)-naphthalenone
Formula:
C₁₀H₁₀O Boiling Pt: 257 °C Density: 1.10 (20 °C) Flash Pt: 129 °C |
MDL Number:
MFCD00001688 CAS Number: 529-34-0 EINECS: 208-460-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...