(±)-Tetrahydrofurfuryl alcohol ≥98.0%
Supplier: TCI America
|
Warning
|
T0106-25ML
T0106-500ML
TCT0106-025MLEA
34.76
CAD
TCT0106-025ML
TCT0106-500ML
(±)-Tetrahydrofurfuryl alcohol ≥98.0%
(±)-Tetrahydrofurfuryl alcohol
CAS Number: 97-99-4
MDL Number: MFCD00005372
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 176
Melting point (°C): -80
Flash Point (°C): 75
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: THFA
Formula:
C₅H₁₀O₂ Boiling Pt: 176 °C Melting Pt: –80 °C Density: 1.05 (20 °C) Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005372 CAS Number: 97-99-4 EINECS: 202-625-6 Merck Index: 12,09353 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...