Tetraethylene glycol ≥95.0%

Supplier: TCI America

Synonyms: 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol

T0099-25G T0099-500G
TCT0099-025GEA 27.26 CAD
TCT0099-025G TCT0099-500G
Tetraethylene glycol ≥95.0%
Tetraethylene glycol

CAS Number: 112-60-7
MDL Number: MFCD00002879
Molecular Formula: C8H18O5
Molecular Weight: 194.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 328
Flash Point (°C): 194
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: Tetraglycol

Formula: C₈H₁₈O₅
MW: 194.23 g/mol
Boiling Pt: 328 °C
Density: 1.13 (20 °C)
Flash Pt: 194 °C
MDL Number: MFCD00002879
CAS Number: 112-60-7
EINECS: 203-989-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47b470a60630e227ee7e0451cba221f282f1d6ec&vid=47b470a60630e227ee7e0451cba221f282f1d6ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR