Tetraethylammonium bromide ≥98.0%

Supplier: TCI America

Synonyms: Tetrylammonium bromide

T0094-25G T0094-500G
TCT0094-025GEA 28.44 CAD
TCT0094-025G TCT0094-500G
Tetraethylammonium bromide ≥98.0%
Tetraethylammonium bromide

CAS Number: 71-91-0
MDL Number: MFCD00011825
Molecular Formula: C8H20BrN
Molecular Weight: 210.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 285

Formula: C₈H₂₀BrN
MW: 210.16 g/mol
Melting Pt: 285 °C
Density: 1.43 g/cm³ (20 °C)
MDL Number: MFCD00011825
CAS Number: 71-91-0
EINECS: 200-769-4
Merck Index: 13,09274
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13ed904e97bcdb01f1a71c12d219d6fe202c1391&vid=13ed904e97bcdb01f1a71c12d219d6fe202c1391&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR