3,4,5-Trimethoxybenzaldehyde ≥98.0%

Supplier: TCI America

T0059-25G T0059-100G
TCT0059-025GEA 53.72 CAD
TCT0059-025G TCT0059-100G
3,4,5-Trimethoxybenzaldehyde ≥98.0%
3,4,5-Trimethoxybenzaldehyde

CAS Number: 86-81-7
MDL Number: MFCD00003364
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 165
Melting point (°C): 76

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 165 °C
Melting Pt: 76 °C
Density: 1.367 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003364
CAS Number: 86-81-7
EINECS: 201-701-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f25986735ed569294f3b86cf2c6518df72760202&vid=efeaa8d20066716b4da0d4984c46f695b9117e1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR