Tetrabutylammonium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T0055-25G T0055-5G T0055-100G
TCT0055-025GEA 210.66 CAD
TCT0055-025G TCT0055-005G TCT0055-100G
Tetrabutylammonium chloride ≥98.0% (by titrimetric analysis)
Tetrabutylammonium chloride

CAS Number: 1112-67-0
MDL Number: MFCD00011635
Molecular Formula: C16H36ClN
Molecular Weight: 277.92
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 70
Flash Point (°C): 113

Formula: C₁₆H₃₆ClN
Melting Pt: 70 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00011635
CAS Number: 1112-67-0
EINECS: 214-195-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=499cfeab81862d6395b92994506f1baed03e9cba&vid=499cfeab81862d6395b92994506f1baed03e9cba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR