Tetrabutylammonium bromide ≥98.0%

Supplier: TCI America

T0054-500G T0054-25G T0054-100G
TCT0054-500GEA 269.96 CAD
TCT0054-500G TCT0054-025G TCT0054-100G
Tetrabutylammonium bromide ≥98.0%
Tetrabutylammonium bromide

CAS Number: 1643-19-2
MDL Number: MFCD00011633
Molecular Formula: C16H36BrN
Molecular Weight: 322.38
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 103

Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 103 °C
Density: 1.148 g/cm³ (20 °C)
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4767b5a29bd91e6d19915e4460d8e1123f8874b&vid=d4767b5a29bd91e6d19915e4460d8e1123f8874b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR