Tetrabromophthalic anhydride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

T0046-25G T0046-100G
TCT0046-025GEA 30.02 CAD
TCT0046-025G TCT0046-100G
Tetrabromophthalic anhydride ≥95.0% (by titrimetric analysis)
Tetrabromophthalic anhydride

CAS Number: 632-79-1
MDL Number: MFCD00005919
Molecular Formula: C8Br4O3
Molecular Weight: 463.70
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 280

Formula: C₈Br₄O₃
MW: 463.7 g/mol
Boiling Pt: 477 °C (1013 hPa)
Melting Pt: 280 °C
Density: 2.93 g/cm³ (20 °C)
MDL Number: MFCD00005919
CAS Number: 632-79-1
EINECS: 211-185-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26d21f53485cd599e793df00fae1a327486aa9cc&vid=26d21f53485cd599e793df00fae1a327486aa9cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR