1,2,4,5-Tetrabromobenzene ≥97.0%

Supplier: TCI America
Warning

T0031-5G T0031-25G
TCT0031-005GEA 91.64 CAD
TCT0031-005G TCT0031-025G
1,2,4,5-Tetrabromobenzene ≥97.0%
1,2,4,5-Tetrabromobenzene

CAS Number: 636-28-2
MDL Number: MFCD00000063
Molecular Formula: C6H2Br4
Molecular Weight: 393.70
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 180

Formula: C₆H₂Br₄
MW: 393.7 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00000063
CAS Number: 636-28-2
EINECS: 211-253-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc2bb335ef090077291f440d6ba0f593307381e9&vid=24cbff081857a08e78b964928569668e2d061ebb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR