(±)-α-Terpineol ≥80.0%

Supplier: TCI America
Warning

T0022-25ML T0022-500ML
TCT0022-025MLEA 35.38 CAD
TCT0022-025ML TCT0022-500ML
(±)-α-Terpineol ≥80.0%
(±)-α-Terpineol

CAS Number: 98-55-5
MDL Number: MFCD00001557
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 101
Specific Gravity (20/20): 0.94


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: 1-p-Menthen-8-ol

Formula: C₁₀H₁₈O
Density: 0.94 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a2ae161197eb313a5f7642db02ac8ea0b2557bf&vid=8a2ae161197eb313a5f7642db02ac8ea0b2557bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR