p-Terphenyl ≥99.0%

Supplier: TCI America

T0020-25G T0020-500G T0020-100G
TCT0020-025GEA 75.57 CAD
TCT0020-025G TCT0020-500G TCT0020-100G
p-Terphenyl ≥99.0%
p-Terphenyl

CAS Number: 92-94-4
MDL Number: MFCD00003061
Molecular Formula: C18H14
Molecular Weight: 230.31
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 389
Melting point (°C): 211
Flash Point (°C): 207
Lambda max.: 276 nm (Cyclohexane)


Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials), Analytical Science, Scintillators
Other Product Names / Synonyms: 1,4-Diphenylbenzene

Formula: C₁₈H₁₄
Boiling Pt: 389 °C
Melting Pt: 211 °C
Density: 1.234 g/cm³ (0 °C)
Flash Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00003061
CAS Number: 92-94-4
EINECS: 202-205-2
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb50149662699269df3fdb150afd26bb425ba068&vid=fb50149662699269df3fdb150afd26bb425ba068&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR