Terephthalonitrile ≥98.0%
Supplier: TCI America
Synonyms:
1,4-Benzenedicarbonitrile, Terephthalodinitrile, Benzene-1,4-dicarbonitrile, 1,4-Dicyanobenzene
T0016-500G
T0016-25G
T0016-100G
TCT0016-500GEA
541.94
CAD
TCT0016-500G
TCT0016-025G
TCT0016-100G
Terephthalonitrile ≥98.0%
Terephthalonitrile
CAS Number: 623-26-7
MDL Number: MFCD00001810
Molecular Formula: C8H4N2
Molecular Weight: 128.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 228
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: 1,4-Dicyanobenzene
Formula:
C₈H₄N₂ MW: 128.13 g/mol Melting Pt: 228 °C Density: 1.29 g/cm³ (20 °C) |
MDL Number:
MFCD00001810 CAS Number: 623-26-7 EINECS: 210-783-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...