Dimethyl terephthalate ≥99.0%

Supplier: TCI America

T0015-25G T0015-500G
TCT0015-025GEA 54.53 CAD
TCT0015-025G TCT0015-500G
Dimethyl terephthalate ≥99.0%
Dimethyl terephthalate

CAS Number: 120-61-6
MDL Number: MFCD00008440
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 142
Flash Point (°C): 153


Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Terephthalic Acid Dimethyl Ester

Formula: C₁₀H₁₀O₄
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: 142 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00008440
CAS Number: 120-61-6
EINECS: 204-411-8
Merck Index: 12,09306
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dff0c7dc19e9750cc68ac4de47bf523299de6967&vid=dff0c7dc19e9750cc68ac4de47bf523299de6967&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR