Diethyl terephthalate ≥99.0%

Supplier: TCI America

T0014-25G T0014-500G
TCT0014-025GEA 94.86 CAD
TCT0014-025G TCT0014-500G
Diethyl terephthalate ≥99.0%
Diethyl terephthalate

CAS Number: 636-09-9
MDL Number: MFCD00039891
Molecular Formula: C12H14O4
Molecular Weight: 222.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 142
Melting point (°C): 45
Flash Point (°C): 152


Other Product Names / Synonyms: Terephthalic Acid Diethyl Ester

Formula: C₁₂H₁₄O₄
Boiling Pt: 142 °C
Melting Pt: 45 °C
Flash Pt: 152 °C
MDL Number: MFCD00039891
CAS Number: 636-09-9
EINECS: 211-249-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0d6c8134889d63af518e9364560504991edff7d&vid=f0d6c8134889d63af518e9364560504991edff7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR