4-Formylbenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0011-25G T0011-500G T0011-100G
TCT0011-025GEA 97.22 CAD
TCT0011-025G TCT0011-500G TCT0011-100G
4-Formylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Formylbenzoic acid

CAS Number: 619-66-9
MDL Number: MFCD00006951
Molecular Formula: C8H6O3
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 256
Lambda max.: 298 nm (Hexane)


Other Product Names / Synonyms: 4-Carboxybenzaldehyde, 4-Formylbenzoic Acid

Formula: C₈H₆O₃
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 256 °C
MDL Number: MFCD00006951
CAS Number: 619-66-9
EINECS: 210-607-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4afaee977ee451a2cc2afe7e876286ee53a5e7a&vid=f4afaee977ee451a2cc2afe7e876286ee53a5e7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR