L(+)-Diethyl tartrate ≥98.0%

Supplier: TCI America

Synonyms: (+)-Diethyl L-tartrate, (R,R)-(+)-Tartaric acid diethyl ester, (2R,3R)-(+)-Diethyl tartrate, Diethyl L-tartrate, L(+)-Tartaric acid diethyl ester, (2R,3R)-Diethyl 2,3-dihydroxysuccinate, (R,R)-(+)-Diethyl tartrate

T0003-500G T0003-25G T0003-100G
TCT0003-500GEA 451.88 CAD
TCT0003-500G TCT0003-025G TCT0003-100G
L(+)-Diethyl tartrate ≥98.0%
L(+)-Diethyl tartrate

CAS Number: 87-91-2
MDL Number: MFCD00009143
Molecular Formula: C8H14O6
Molecular Weight: 206.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 162
Flash Point (°C): 152
Specific Gravity (20/20): 1.21
Specific rotation [a]20/D: 7.8 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(+)-Tartaric Acid Diethyl Ester

Formula: C₈H₁₄O₆
MW: 206.2 g/mol
Boiling Pt: 162 °C
Melting Pt: 18.7 °C
Density: 1.21 (20 °C)
Flash Pt: 152 °C
MDL Number: MFCD00009143
CAS Number: 87-91-2
EINECS: 201-783-3
Merck Index: 13,03886
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11dd299b548c40e754d0af1546add7685576bbbf&vid=11dd299b548c40e754d0af1546add7685576bbbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR